ORCA Response Temperature-Controlled Packaging Solutions
À propos de cet article
ORCA Response is a multi-use, handheld, temperature-controlled packaging solution for transporting smaller payloads of red blood cells, platelets, pharmaceutical products and be easily carried and stored, making it the ideal solution for taking personal medicines on long journeys, including suitability as hand luggage on aeroplanes.
Easy to assemble
Utilises Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies
Universal pack out, Simple conditioning and assembly
3 PCM temperature ranges: 2...8 ˚C, 15...25 ˚C, –20 ˚C
Qualified for use with dry ice as coolant
Optional carry bag for easy portability
Optional ORCA Medic carry bag for military applications
Suitable as hand luggage when flying
Advanced Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies ORCA response is qualified against ISTA 7D Summer and Winter temperature profiles, and enable 60+ hours of temperature protection.
Designed with the end user in mind ORCA Response combines high-performance technology and ease of use where rapid response packaging is required. This optimum performance and small volumetric weight make ORCA Response a clear choice for temperature-sensitive transportation and emergency applications.
3 Options disponibles ci-dessous
Product Details & Documents
ORCA Response is a multi-use, handheld, temperature-controlled packaging solution for transporting smaller payloads of red blood cells, platelets, pharmaceutical products and be easily carried and stored, making it the ideal solution for taking personal medicines on long journeys, including suitability as hand luggage on aeroplanes.
Easy to assemble
Utilises Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies
Universal pack out, Simple conditioning and assembly
3 PCM temperature ranges: 2...8 ˚C, 15...25 ˚C, –20 ˚C
Qualified for use with dry ice as coolant
Optional carry bag for easy portability
Optional ORCA Medic carry bag for military applications
Suitable as hand luggage when flying
Advanced Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies ORCA response is qualified against ISTA 7D Summer and Winter temperature profiles, and enable 60+ hours of temperature protection.
Designed with the end user in mind ORCA Response combines high-performance technology and ease of use where rapid response packaging is required. This optimum performance and small volumetric weight make ORCA Response a clear choice for temperature-sensitive transportation and emergency applications.
Product Family Options
ORCA Response
Informations sur le produit
- W×D×H ext.DisponibilitéPrix
- DGINORS200DIORCA Response 2 L dry ice, 7,7 L payload volume, 1,9 kgORCA Response 2 L dry ice, 7,7 L payload volume, 1,9 kg347×260×223 mmConnexion Pour la disponibilité1 * 1 ST165,00 €Spécifications :New ProductNew Product
- DGINORS223ORCA Response 2 L , –20 °C, 2,2 L payload volume, 6 kgORCA Response 2 L , –20 °C, 2,2 L payload volume, 6 kg347×260×223 mmConnexion Pour la disponibilité1 * 1 ST267,00 €Spécifications :New ProductNew Product
ORCA Response Carry Bag
Informations sur le produit
- W×D×H ext.DisponibilitéPrix
- DGINORSBAG2LORCA Response 2 L carry bag, with carry strapORCA Response 2 L carry bag, with carry strap347×273×233 mmConnexion Pour la disponibilité1 * 1 ST93,80 €Spécifications :New ProductNew Product
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.