Order Entry
Belgium
ContactUsLinkComponent
Streptococcus pyogenes Recombinant Cas9 NC-NLS (from E. coli)
Streptococcus pyogenes Recombinant Cas9 NC-NLS (from E. coli)
Catalog # ORIGTP790148
Supplier:  OriGene
New Product
CAS Number:  
Streptococcus pyogenes Recombinant Cas9 NC-NLS (from E. coli)
Catalog # ORIGTP790148
Supplier:  OriGene
New Product
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    S. pyogenes
  • Tag sequence:
    N-His
  • Storage conditions:
    Store at −20 °C
  • Endotoxin content:
    < 0,5 EU per 1 µg of the protein by the LAL
  • Biological activity:
    In vitro digestion of a DNA fragment with the recombinant S.pyrogene Cas9 nuclease (Cat# TP790148). The input DNA is XmnI-linearized vector (PS100074) was cleaved by with a synthetic sgRNA
  • Protein/peptide name:
    Cas9 NC-NLS
  • Purity:
    >90% as determined by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    165,0 kDa
  • Sequence:
    1 MHHHHHHENL YFQGAIPKKK RKVEDPKKKR KVDIAMDKKY SIGLDIGTNS VGWAVITDEY 61 KVPSKKFKVL GNTDRHSIKK NLIGALLFDS GETAEATRLK RTARRRYTRR KNRICYLQEI 121 FSNEMAKVDD SFFHRLEESF LVEEDKKHER HPIFGNIVDE VAYHEKYPTI YHLRKKLVDS 181 TDKADLRLIY LALAHMIKFR GHFLIEGDLN PDNSDVDKLF IQLVQTYNQL FEENPINASG 241 VDAKAILSAR LSKSRRLENL IAQLPGEKKN GLFGNLIALS LGLTPNFKSN FDLAEDAKLQ 301 LSKDTYDDDL DNLLAQIGDQ YADLFLAAKN LSDAILLSDI LRVNTEITKA PLSASMIKRY 361 DEHHQDLTLL KALVRQQLPE KYKEIFFDQS KNGYAGYIDG GASQEEFYKF IKPILEKMDG 421 TEELLVKLNR EDLLRKQRTF DNGSIPHQIH LGELHAILRR QEDFYPFLKD NREKIEKILT 481 FRIPYYVGPL ARGNSRFAWM TRKSEETITP WNFEEVVDKG ASAQSFIERM TNFDKNLPNE 541 KVLPKHSLLY EYFTVYNELT KVKYVTEGMR KPAFLSGEQK KAIVDLLFKT NRKVTVKQLK 601 EDYFKKIECF DSVEISGVED RFNASLGTYH DLLKIIKDKD FLDNEENEDI LEDIVLTLTL 661 FEDREMIEER LKTYAHLFDD KVMKQLKRRR YTGWGRLSRK LINGIRDKQS GKTILDFLKS 721 DGFANRNFMQ LIHDDSLTFK EDIQKAQVSG QGDSLHEHIA NLAGSPAIKK GILQTVKVVD 781 ELVKVMGRHK PENIVIEMAR ENQTTQKGQK NSRERMKRIE EGIKELGSQI LKEHPVENTQ 841 LQNEKLYLYY LQNGRDMYVD QELDINRLSD YDVDHIVPQS FLKDDSIDNK VLTRSDKNRG 901 KSDNVPSEEV VKKMKNYWRQ LLNAKLITQR KFDNLTKAER GGLSELDKAG FIKRQLVETR 961 QITKHVAQIL DSRMNTKYDE NDKLIREVKV ITLKSKLVSD FRKDFQFYKV REINNYHHAH 1021 DAYLNAVVGT ALIKKYPKLE SEFVYGDYKV YDVRKMIAKS EQEIGKATAK YFFYSNIMNF 1081 FKTEITLANG EIRKRPLIET NGETGEIVWD KGRDFATVRK VLSMPQVNIV KKTEVQTGGF 1141 SKESILPKRN SDKLIARKKD WDPKKYGGFD SPTVAYSVLV VAKVEKGKSK KLKSVKELLG 1201 ITIMERSSFE KNPIDFLEAK GYKEVKKDLI IKLPKYSLFE LENGRKRMLA SAGELQKGNE 1261 LALPSKYVNF LYLASHYEKL KGSPEDNEQK QLFVEQHKHY LDEIIEQISE FSKRVILADA 1321 NLDKVLSAYN KHRDKPIREQ AENIIHLFTL TNLGAPAAFK YFDTTIDRKR YTSTKEVLDA 1381 TLIHQSITGL YETRIDLSQL GGDPKKKRKV EDPKKKRKVD TR
  • Concentration:
    10 µM

Specifications

About this item

Purified recombinant protein of S. pyogenes Cas9 endonuclease (Cas9) containing Simian virus 40 (SV40) T antigen nuclear localisation sequence (NLS) on the N- and C- termini of the protein., with N-terminal HIS tag, expressed in E.coli, sterile-filtered, suitable for cell culture.

  • Shipped in dry ice

Buffer: 10 mM Tris-HCl, pH 7,4, 300 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 50% glycerol

Stability: Stable for 12 months from the date of receipt of the product under proper storage and handling conditions

Caution:

Avoid repeated freeze-thaw cycles. For testing in cell culture applications, please filter before use. Note that you may experience some loss of protein during the filtration process.