Orders LinkContactUsLinkComponent
Combi-Rack for Test Tubes
Combi-Rack for Test Tubes
New Product
Supplier:   NEOLAB
Combi-Rack for Test Tubes
Supplier:  NEOLAB
New Product
Certificates
Combi-Rack for Test Tubes
3 Options Available Below
Product Details & Documents
Combi rack for test tubes, 96-position.
  • Double-sided test tube rack with lid
    Available in 7 colours
    Made of PP
    Autoclavable up to 121 °C
On the upper side there are holes for test tubes with a capacity of 1,5 and 2 ml, on the lower one places for test tubes with a capacity of 1,5 and 2 ml. 0,5 ml.
Product Family Options
Product Information
  • Colour
    Material
    W×D×H
    Availability
    Price
  • NEOL2-2572
    Combi-Rack for test tubes, 96-position
    Combi-Rack for test tubes, 96-position
    Red
    PP
    210×115×28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,40 €
    30,40 €
    1 * 1 ST
       30,40 €
    Specifications:
    New Product
    New Product
  • NEOL2-2573
    Combi-rack for test tubes, 96-position
    Combi-rack for test tubes, 96-position
    Violet
    PP
    210×115×28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,40 €
    30,40 €
    1 * 1 ST
       30,40 €
    Specifications:
    New Product
    New Product
  • NEOL2-2575
    Combi-rack for test tubes, 96-position
    Combi-rack for test tubes, 96-position
    Green
    PP
    210×115×28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,40 €
    30,40 €
    1 * 1 ST
       30,40 €
    Specifications:
    New Product