Order Entry
Belgium
ContactUsLinkComponent
Mag-Bind® Endo-free Plasmid Midi Kit
Mag-Bind® Endo-free Plasmid Midi Kit
Supplier:  OMEGA BIO-TEK
New Product
Certificates
undefined
Mag-Bind® Endo-free Plasmid Midi Kit
Supplier:  OMEGA BIO-TEK
New Product
Limited Time Sale
Select items below are currently on sale

Specifications

  • Prep size:
    Scalable
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Instrument Compatibility:
    MagBinder® Fit24 nucleic acid purification system

Specifications

Frequently Bought Together