ContactUsLinkComponent
Avantor® PUPSIT Assemblies
New Product
undefined
Avantor® PUPSIT Assemblies
New Product
  • Catalog No:
  • 77865-532
  • 77865-530
  • 77865-528
  • Filtration area:
  • 0,6 m²
  • 0,6 m²
  • 0,01 m²
  • Filter:
  • Liquid filter: Sartorius Sartopore 2 / Barrier filter: Zebragard Barrier
  • Liquid filter: Cytiva Supor / Barrier filter: Zebragard Barrier
  • Liquid filter: Millipore Durapore / Barrier filter: Millipak Barrier
  • Sensor:
  • Single-use Gauge Tee (SGT)
  • Single-use Gauge Tee (SGT)
  • Pendotech
  • Tubing:
  • Reinforced silicone
  • Reinforced silicone
  • Reinforced silicone
Frequently Bought Together