Order Entry
Belgium
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Test Kit for Cadmium
NANOCOLOR® Photometric Standard Test Kit for Cadmium
Catalog # MANA918131
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Test Kit for Cadmium
Catalog # MANA918131
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Dithizone
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Cadmium, test no. 1-131
  • Dimensions:
    160×110×130 mm (Including packaging)
  • Model:
    MANA918131
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    18
  • Weight:
    485 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;Advance;500 D
  • Tests per kit:
    25
  • Range:
    0,002 - 0,50 mg/L (Cd²⁺)

Specifications

About this item

Photometric test kit for the determination of cadmium in water samples.

  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II

Sea water analysis: No

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent: Yes

Digestion: Yes

NanOx metal: Yes

NanOx: No

Delivery: Rugged box with reagents for 25 tests.