Order Entry
Belgium
ContactUsLinkComponent
 
NANOCOLOR® Photometric Standard Test Kit for Phenol
  634-8123
 :  
undefined
NANOCOLOR® Photometric Standard Test Kit for Phenol
  634-8123
 :  

 

  • Chemical method:
    4 -Nitroaniline
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Phenol, test no.1-75
  • Dimensions:
    160×110×130 mm (including packaging)
  • Seq:
    0
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    36
  • Weight:
    605,7 g (including packaging)
  • Type:
    Hazardous material: Yes
  • For unit:
    UV/VIS II; VIS II; Advance; 500 D; MT Easy VIS; MT Easy UV
  • Tests per kit:
    500
  • Range:
    0,01 - 7,0 mg/L (Phenol)

 

 

Photometric test kit for the determination of phenols in water samples.

  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

 : Remark: Measuring range on NANOCOLOR VIS II