Order Entry
Belgium
ContactUsLinkComponent
KIMBLE® KIMAX® Cassia Volumetric Flask, Class A, TC, with Pennyhead Glass Stopper
KIMBLE® KIMAX® Cassia Volumetric Flask, Class A, TC, with Pennyhead Glass Stopper
Catalog # SCERSSP28066-110
Supplier:  DWK Life Sciences
KIMBLE® KIMAX® Cassia Volumetric Flask, Class A, TC, with Pennyhead Glass Stopper
Catalog # SCERSSP28066-110
Supplier:  DWK Life Sciences

Specifications

  • Capacity:
    110 ml
  • Height:
    260 mm (including stopper)
  • Tolerance:
    ±0,08 ml

Specifications

About this item

The KIMBLE® KIMAX® Cassia Volumetric Flask used in assay of oils of cinnamon, chenopodium, cloves, spearmint and other essential oils, according to methods of the United States Pharmacopoeia and the Essential Oil Association of the USA.

  • Manufactured from borosilicate glass 3,3

  • Calibrated 'To Contain'
  • Flask neck is graduated from 100 to 110 ml in 0,1 ml subdivisions, with a ring at each 1ml
  • With a marking spot and a duarbale white ceramic enamel scale
  • ASTM Class A

Supplied with pennyhead glass stopper (850100-0013).