Order Entry
Belgium
Orders LinkContactUsLinkComponent
Precision™ Circulating Water Baths

Precision™ Circulating Water Baths

Supplier:   THERMO ELECTRON LED
Precision™ Circulating Water Baths
Certificates
Precision™ Circulating Water Baths

2 Options Available Below

Product Details & Documents

Ensure temperature uniformity and control with these Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow

  • Easily clean and maintain bath with coil-free internal design
  • Optimise scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms

Ordering information: Not for use for IVD/MD in some countries.

Specifications for Product Family

Specifications

  • Catalog No:
  • HAKATSCIR19
  • HAKATSCIR89
  • Capacity:
  • 19 L
  • 89 L
  • Electrical:
  • Global Voltage 100/115 V; 200/230 V; 50/60 Hz
  • Global Voltage 100/115 V; 200/230 V; 50/60 Hz
  • Ext. W×D×H:
  • 305×387×192 mm
  • 483×914×300 mm

Specifications

Product Family Options

hs, Water Baths and Shaking Baths

Product Information

  • Availability
    Price
  • HAKATSCIR19
    Circulating water bath, 19 L
    Circulating water bath, 19 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 910,00 €
    2 910,00 €
    1 * 1 ST
       2 910,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    HAKATSCIR19
    Capacity
    19 L
    Electrical
    Global Voltage 100/115 V; 200/230 V; 50/60 Hz
    Ext. W×D×H
    305×387×192 mm
  • HAKATSCIR89
    Circulating water bath, 89 L
    Circulating water bath, 89 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 430,00 €
    4 430,00 €
    1 * 1 ST
       4 430,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    HAKATSCIR89
    Capacity
    89 L
    Electrical
    Global Voltage 100/115 V; 200/230 V; 50/60 Hz
    Ext. W×D×H
    483×914×300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.