ContactUsLinkComponent
b.safe Assembly Aid for Desktop Mount
b.safe Assembly Aid for Desktop Mount
Catalog # BOHLR1103-01
Supplier:  Bohlender
New Product
undefined
b.safe Assembly Aid for Desktop Mount
Catalog # BOHLR1103-01
Supplier:  Bohlender
New Product
Specifications
  • Description:
    Assembly aid for desktop mount
Specifications
About this item
Two-part assembly aid for b.safe desktop mounts GL 45, Cat. No. BOHLR1100-45 and BOHLR1100-12.
  • Top connection: female thread GL 45
  • Bottom connection: inside hexagon SW38
  • Made of PP
Remove the protective film from the adhesive pad of the desktop mount and place it in the hole provided in the worktop. Screw the upper part of the assembly aid onto the GL 45 connection of the desktop mount. Then screw the lock nut from below onto the thread provided as far up as possible against the worktop. To tighten the desktop mount, push the lower part of the assembly aid (inside hexagon) onto the desktop mount from below.
Screwdrivers, for example, can be inserted into the holes in the top and bottom part to hold the assembly aid at the top and tighten the lock nut at the bottom.