Human Recombinant PFAS (from E. coli)
Product Details & Documents
Recombinant Human PFAS protein (Tagged) is a Human Fragment protein, in the 1064 to 1302 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Form:Liquid
- Protein/peptide name:PFAS
- Protein synonyms:KIAA0361|Phosphoribosylformylglycinamidine synthase|FGAM synthase|FGAMS|Formylglycinamide ribonucleotide amidotransferase|Formylglycinamide ribotide amidotransferase|Phosphoribosylformylglycineamide amidotransferase|FGAR amidotransferase|FGAR-AT
- Purity:>85%
- Source:E. coli
- Biological activity:Standard
- Shipping temperature:20
- Cat. No.:ABCAAB236198-100
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C
- Formulation:pH: 7,2 - 7,4, Constituents: Tris buffer, 50% Glycerol (glycerin, glycerine)
- Conjugation:Unconjugated
- Tested applications:SDS-PAGE
- Species:Human
- Animal-free:N
Specifications
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



