Order Entry
Belgium
Orders LinkContactUsLinkComponent
Human Recombinant PRRT2 (from E. coli)

Human Recombinant PRRT2 (from E. coli)

Catalog # ABCAAB177630-50
Supplier:   Abcam
Human Recombinant PRRT2 (from E. coli)
Catalog # ABCAAB177630-50
Supplier:  Abcam
CAS Number:  
Human Recombinant PRRT2 (from E. coli)
Human Recombinant PRRT2 (from E. coli)
CAS Number:  
Price
544,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Recombinant human PRRT2 protein is a Human Fragment protein, in the 1 to 268 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    PRRT2
  • Protein synonyms:
    Proline-rich transmembrane protein 2|Dispanin subfamily B member 3|DSPB3|PRRT2
  • Purity:
    >85%
  • Concentration:
    0,25 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB177630-50
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,88% Sodium chloride, 0,32% Tris-HCl buffer, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Conjugation:
    Unconjugated
  • Tested applications:
    MS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.