Wash Buffer Concentrate (20x)
Catalog # ABCAAB172375-30
Supplier: Abcam
CAS Number:
Specifications
- Description:Wash buffer concentrate (20X)
- Form:Pre-coated
- Storage temperature:4 °C
- Volume:30 ml
Specifications
About this item
Wash Buffer Concentrate (20X) (ab172375) is part of the reagents, controls & accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.