Order Entry
Belgium
ContactUsLinkComponent
Rat Interferon gamma ELISPOT Set
Rat Interferon gamma ELISPOT Set
Supplier:  Abcam
Certificates
undefined
Rat Interferon gamma ELISPOT Set
Supplier:  Abcam

Specifications

  • Species reactivity:
    Rat
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Antigen name:
    Interferon gamma
  • Storage temperature:
    +4 ℃
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO

Specifications

Frequently Bought Together