Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample Diluent NBP

Sample Diluent NBP

Sample diluent NBP, Research use only (RUO), 20 ml
Catalog # ABCAAB221826-20
Supplier:   Abcam
Sample Diluent NBP
Catalog # ABCAAB221826-20
Supplier:  Abcam
CAS Number:  
Sample Diluent NBP
Sample Diluent NBP

Sample diluent NBP, Research use only (RUO), 20 ml

CAS Number:  
Price
82,00 €
Log in for availability
1 * 20 mL
Product Details & Documents

Sample Diluent NBP is part of the reagents, controls and accessories range.

Caution: RUO.

Specifications

Specifications

  • Description:
    Sample diluent NBP
  • Type:
    Research use only (RUO)
  • Volume:
    20 ml
  • Cat. No.:
    ABCAAB221826-20
  • Storage temperature:
    4 °C

Specifications