Order Entry
Belgium
ContactUsLinkComponent
Complex I Enzyme Activity Dipstick Assay Kit
Complex I Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109720-48
Supplier:  Abcam
CAS Number:  
undefined
Complex I Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109720-48
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Complex I Enzyme activity dipstick assay kit
  • Protein:
    Complex I
  • Species reactivity:
    Other, Human, Mouse, Rat
  • Sample type:
    Cell culture extracts, Tissue, Cell culture supernatant
  • Storage temperature:
    +4 ℃
  • Processing time:
    Multiple steps
  • Product type:
    Pre-coated
  • Tests per kit:
    48 tests

Specifications

About this item

Complex I Enzyme Activity Dipstick Assay kit is an immunoassay kit for the detection of Complex I Enzyme Activity in Mouse, Rat, Cow, Human in cell culture extracts, tissue, cell culture supernatant samples.

Caution: RUO