Mouse Galectin-1 ELISA Kit
Mouse Galectin-1 ELISA kit, 96 tests
ABCAAB253212-1X96
: Abcam
Mouse Galectin-1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Galectin-1 with a sensitivity of 0,17 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
- Description:Mouse Galectin-1 ELISA kit
- Tests per kit:96 tests
- Primary antibody reactivity:Other|Mouse
- Sample type:Heparin plasma, citrate plasma, serum
- Detection range:0,19 - 12 ng/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB253212-1X96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:0,17 ng/ml
- Format:Pre-coated
- Target protein:Galectin 1
- Regulatory status:RUO
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



