Human TNF alpha ELISPOT Kit (with un-coated plates)
: Abcam
Human TNF alpha ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human TNF alpha production in a single cell suspension.
- Species reactivity:Human, Monkey
- Sample type:Suspension cells
- Form:Uncoated
- Storage temperature:4 °C
- Processing time:Multiple steps
- Antigen symbol:TNF alpha
- Regulatory status:RUO
Frequently Bought Together