Order Entry
Belgium
ContactUsLinkComponent
Complex IV Rodent Enzyme Activity Dipstick Assay Kit
Complex IV Rodent Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109878-30
Supplier:  Abcam
CAS Number:  
undefined
Complex IV Rodent Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109878-30
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Complex IV rodent enzyme activity dipstick assay kit
  • Method:
    Sandwich
  • Protein:
    Complex IV
  • Species reactivity:
    Mouse;Rat
  • Sample type:
    Cell culture extracts, tissue
  • Storage temperature:
    Dual temperature (Dry ice)
  • Processing time:
    Multiple steps
  • Product type:
    Pre-coated
  • Tests per kit:
    30 tests

Specifications

About this item

Complex IV Rodent Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Complex IV Rodent Enzyme Activity in Mouse, Rat in Cell culture extracts, Tissue samples.

Certifications: RUO