Mouse Factor D ELISA Kit
Mouse Factor D ELISA kit, 96 tests
Catalog # ABCAAB170244-96
Supplier: Abcam
Jump to:
Product Details & Documents
Mouse Factor D ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Factor D in Mouse in Urine, plasma, Serum samples.
Specifications
Specifications
- Description:Mouse Factor D ELISA kit
- Tests per kit:96 tests
- Primary antibody reactivity:Mouse
- Sample type:Urine, plasma, serum
- Detection range:31,25 - 2000 pg/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:4 h 0 min
- Cat. No.:ABCAAB170244-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:17 pg/ml
- Format:Pre-coated
- Target protein:Factor D/CFD
- Regulatory status:RUO
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



