Order Entry
Belgium
ContactUsLinkComponent
Rat Interferon gamma ELISPOT Kit
Rat Interferon gamma ELISPOT Kit
Supplier:  Abcam
Certificates
undefined
Rat Interferon gamma ELISPOT Kit
Supplier:  Abcam

Specifications

  • Species reactivity:
    Rat
  • Sample type:
    Suspension cells
  • Antigen name:
    Interferon gamma
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Product type:
    Pre-coated

Specifications

Frequently Bought Together