Phosphotyrosine STAT3 ELISA Kit
ABCAAB279941-1X96
: Abcam
:
- Description:Phosphotyrosine STAT3 ELISA Kit
- Tests per kit:96 tests
Phosphotyrosine STAT3 ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine STAT3 in Human in Cell Lysate samples.