Order Entry
Belgium
ContactUsLinkComponent
Mouse IL-2 ELISPOT Set
Mouse IL-2 ELISPOT Set
Supplier:  Abcam
Certificates
undefined
Mouse IL-2 ELISPOT Set
Supplier:  Abcam

Specifications

  • Species reactivity:
    Mouse
  • Sample type:
    Suspension cells
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Antigen symbol:
    IL-2
  • Product type:
    Uncoated

Specifications

Frequently Bought Together