Phosphotyrosine ZAP70 ELISA Kit
Catalog # ABCAAB279970-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Phosphotyrosine ZAP70 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Phosphotyrosine ZAP70 ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine ZAP70 in Human in cell lysate samples.