Human CD5 ELISA Kit - Extracellular
Human CD5 ELISA kit - Extracellular, 96 tests
Catalog # ABCAAB314853-96TST
New Product
Supplier: Abcam
Human CD5 ELISA Kit - Extracellular
Catalog # ABCAAB314853-96TST
Supplier: Abcam
New Product
CAS Number:
Jump to:
Product Details & Documents
Human CD5 ELISA kit - Extracellular is a single-wash 90-min Simplestep used to quantify Human CD5 with a sensitivity of 2,131 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.
Specifications
Specifications
- Description:Human CD5 ELISA kit - Extracellular
- Tests per kit:96 tests
- Primary antibody reactivity:Human
- Sample type:Serum, Heparin Plasma, Citrate plasma, EDTA Plasma, Cell culture supernatant
- Detection range:10,156 - 650 pg/ml
- Storage temperature:4 ℃
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB314853-96TST
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:2,131 pg/ml
- Format:Pre-coated
- Target protein:CD5
- Regulatory status:RUO
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



