Phosphotyrosine CSK ELISA Kit
ABCAAB279765-1X96
: Abcam
:
- Description:Phosphotyrosine CSK ELISA Kit
- Tests per kit:96 tests
Phosphotyrosine CSK ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine CSK in Human in Cell Lysate samples.