Orders LinkContactUsLinkComponent
Immunoassay Blocking Buffer
Immunoassay Blocking Buffer
Marketsource
Supplier:   Abcam
Immunoassay Blocking Buffer
Supplier:  Abcam
Marketsource
Certificates
Immunoassay Blocking Buffer
2 Options Available Below
Product Details & Documents
Immunoassay blocking buffer effectively preserves the conformation and activity of dried proteins in immunoassays.
Caution: RUO.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • ABCAAB171534-50
  • ABCAAB171534-1000
  • Storage temperature:
  • Store at room temperature
  • Store at room temperature
Specifications
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • ABCAAB171534-50
    Immunoassay blocking buffer, pre-coated
    Immunoassay blocking buffer, pre-coated
    50 ml
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 82,00 €
    82,00 €
    1 * 50 mL
       82,00 €
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB171534-50
    Storage temperature
    Store at room temperature
    Marketsource
  • ABCAAB171534-1000
    Immunoassay blocking buffer, pre-coated
    Immunoassay blocking buffer, pre-coated
    1000 ml
    Log in for availability
    Log in for availability
    1 * 1.000 mL
    1 * 1.000 mL - 431,00 €
    431,00 €
    1 * 1.000 mL
       431,00 €
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB171534-1000
    Storage temperature
    Store at room temperature
    Marketsource