Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rat Complement C3 ELISA Kit

Rat Complement C3 ELISA Kit

Rat Complement C3 ELISA kit, 96 tests
Catalog # ABCAAB157731-96
Supplier:   Abcam
Rat Complement C3 ELISA Kit
Catalog # ABCAAB157731-96
Supplier:  Abcam
CAS Number:  
Rat Complement C3 ELISA Kit
Rat Complement C3 ELISA Kit

Rat Complement C3 ELISA kit, 96 tests

CAS Number:  
Price
790,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Rat Complement C3 ELISA Kit ab157731 is used to quantify Rat Complement C3 with a sensitivity of 15,8 pg/ml. Sandwich ELISA Kit.

Specifications

Specifications

  • Description:
    Rat Complement C3 ELISA kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB157731-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    2,82 ng/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Sample type:
    Plasma, serum
  • Target protein:
    C3/C3b
  • Detection range:
    12,5 - 800 ng/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.