Phosphotyrosine STAT5 ELISA Kit
ABCAAB279945-1X96
: Abcam
:
- Description:Phosphotyrosine STAT5 ELISA kit
- Tests per kit:96 tests
Phosphotyrosine STAT5 ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine STAT5 in Human in cell lysate samples.