Immunoassay Blocking (BSA Free)
Supplier: Abcam
Certificates
About this item
Immunoassay Blocking (BSA Free) (ab171535) maintains the activity of antibodies and other biomolecules adsorbed or immobilized to solid substrates.
Specifications
- Storage temperature:Store at room temperature