Order Entry
Belgium
ContactUsLinkComponent
TMB substrate 1X
TMB substrate 1X
Catalog # ABCAAB210902-120
Supplier:  Abcam
CAS Number:  
undefined
TMB substrate 1X
Catalog # ABCAAB210902-120
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    TMB substrate 1X
  • Size:
    120 ml
  • Storage temperature:
    4 °C
  • Plate types:
    Uncoated
  • Processing volume:
    Multiple steps

Specifications

About this item

TMB substrate recommended for use with the Matched Antibody Pair kits for sandwich ELISA.