Human Complement C3b ELISA Kit
Human Complement C3b ELISA kit, 96 tests
Catalog # ABCAAB195461-96
Supplier: Abcam
Jump to:
Product Details & Documents
Human Complement C3b ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement C3b in Human in Cerebral Spinal Fluid, Saliva, Urine, Plasma, Milk, Serum samples.
Specifications
Specifications
- Description:Human Complement C3b ELISA kit
- Tests per kit:96 tests
- Primary antibody reactivity:Human
- Sample type:Cerebral Spinal Fluid, Saliva, Urine, Plasma, Milk, Serum
- Detection range:0,3 - 1,1 ng/ml
- Storage temperature:+4 °C
- Assay Type:Sandwich
- Time to Results:4 h 0 min
- Cat. No.:ABCAAB195461-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:67 pg/ml
- Format:Pre-coated
- Target protein:C3 / C3b
- Regulatory status:RUO
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



