ContactUsLinkComponent
Phosphotyrosine FER ELISA Kit
Catalog # ABCAAB279796-1X96
Supplier:  Abcam
CAS Number:  
undefined
Phosphotyrosine FER ELISA Kit
Catalog # ABCAAB279796-1X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Phosphotyrosine FER ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Phosphotyrosine FER ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine FER in Human in Cell Lysate samples.