Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Guinea Pig Complement C3 ELISA Kit

Guinea Pig Complement C3 ELISA Kit, 96 tests
  ABCAAB157705-96
 :   Abcam
Guinea Pig Complement C3 ELISA Kit
  ABCAAB157705-96
 :  Abcam
 :  
Guinea Pig Complement C3 ELISA Kit
Guinea Pig Complement C3 ELISA Kit

Guinea Pig Complement C3 ELISA Kit, 96 tests

 :  
 
790,00 €
   
1 * 96 Tests
 

Guinea Pig Complement C3 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Guinea Pig Complement C3 in Guinea pig in serum samples.

 

 

  • Description:
    Guinea Pig Complement C3 ELISA Kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB157705-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    1,3083 ng/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Guinea pig
  • Sample type:
    Serum
  • Target protein:
    C3/C3b
  • Detection range:
    3,13 - 3260 ng/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich