Order Entry
Belgium
ContactUsLinkComponent
 
Human Interferon gamma ELISPOT Set
 :  Abcam
 
undefined
Human Interferon gamma ELISPOT Set
 :  Abcam

 

  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Antigen name:
    Interferon gamma
  • Storage temperature:
    +4 ℃
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO

 

Frequently Bought Together