Order Entry
Belgium
ContactUsLinkComponent
Pyruvate dehydrogenase (PDH) Enzyme Activity Dipstick Assay Kit
Pyruvate dehydrogenase (PDH) Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109882-30
Supplier:  Abcam
CAS Number:  
undefined
Pyruvate dehydrogenase (PDH) Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109882-30
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Pyruvate dehydrogenase (PDH) enzyme activity dipstick assay kit
  • Method:
    Sandwich
  • Protein:
    Pyruvate dehydrogenase
  • Species reactivity:
    OTHER, Human, Mouse, Pig, Rat
  • Sample type:
    Cell culture extracts, Tissue
  • Storage temperature:
    Dual Temp (Dry Ice)
  • Processing time:
    Multiple steps
  • Product type:
    Pre-coated
  • Tests per kit:
    30

Specifications

About this item

Pyruvate dehydrogenase (PDH) Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Pyruvate dehydrogenase (PDH) Enzyme Activity in Mouse, Rat, Cow, Human, Pig in Cell culture extracts, Tissue samples.