Feline IL-8 ELISA Kit
Catalog # ABCAAB193688-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Feline IL-8 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Feline IL-8 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Feline IL-8 in Cat in Plasma, Cell culture supernatant, Serum samples.