Order Entry
Belgium
Orders LinkContactUsLinkComponent
Human CBS ELISA Kit

Human CBS ELISA Kit

Human CBS ELISA kit, 96 tests
Catalog # ABCAAB316900-96TST
New Product
Supplier:   Abcam
Human CBS ELISA Kit
Catalog # ABCAAB316900-96TST
Supplier:  Abcam
New Product
CAS Number:  
Human CBS ELISA Kit
Human CBS ELISA Kit

Human CBS ELISA kit, 96 tests

CAS Number:  
Price
774,00 €
Log in for availability
1 * 1 Tests
Product Details & Documents

Human CBS ELISA kit is a single-wash 90-min Simplestep used to quantify Human CBS with a sensitivity of 69.062 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human CBS ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell lysate
  • Detection range:
    187,5 - 12000 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB316900-96TST
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    69,062 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    CBS
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.