ContactUsLinkComponent
Mouse Aspartate Aminotransferase ELISA Kit
Mouse Aspartate Aminotransferase ELISA Kit
Catalog # ABCAAB285300-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse Aspartate Aminotransferase ELISA Kit
Catalog # ABCAAB285300-96
Supplier:  Abcam
CAS Number:  
About this item
Mouse Aspartate Aminotransferase ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Aspartate Aminotransferase in plasma, Tissue Homogenate, serum samples.