Mouse RGM-C ELISA Kit
Catalog # ABCAAB243685-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse RGM-C ELISA kit
Specifications
About this item
Mouse RGM-C ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse RGM-C with a sensitivity of 16.3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.