Order Entry
Belgium
Orders LinkContactUsLinkComponent
Human FGF basic ELISA Kit (FGF2)

Human FGF basic ELISA Kit (FGF2)

Human FGF basic ELISA kit (FGF2), 96 tests
Catalog # ABCAAB99980-96
Supplier:   Abcam
Human FGF basic ELISA Kit (FGF2)
Catalog # ABCAAB99980-96
Supplier:  Abcam
CAS Number:  
Human FGF basic ELISA Kit (FGF2)
Human FGF basic ELISA Kit (FGF2)

Human FGF basic ELISA kit (FGF2), 96 tests

CAS Number:  
Price
836,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human FGF basic ELISA kit (FGF2) is a sandwich (quantitative) ELISA kit for the measurement of Human FGF basic (FGF2) in Human in cell culture extracts, tissue extracts samples.

Specifications

Specifications

  • Description:
    Human FGF basic ELISA kit (FGF2)
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB99980-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    50 pg/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell culture extracts, Tissue Extracts
  • Target protein:
    FGF basic
  • Detection range:
    102,4 - 10000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications