ContactUsLinkComponent
Mouse Fibrinogen ELISA Kit
Mouse Fibrinogen ELISA Kit
  ABCAAB108844-96
 :  Abcam
 :  
undefined
Mouse Fibrinogen ELISA Kit
  ABCAAB108844-96
 :  Abcam
 :  
  • Description:
    Mouse Fibrinogen ELISA kit
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests
Mouse Fibrinogen ELISA kit is a Competitive ELISA kit for the measurement of Mouse Fibrinogen in Mouse in plasma samples.