Oxytocin ELISA Kit
Catalog # ABCAAB133050-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Oxytocin ELISA kit
Specifications
About this item
Oxytocin ELISA kit is a Competitive ELISA kit for the measurement of Oxytocin in Human in Cerebral Spinal Fluid, Saliva, urine, plasma, cell culture supernatant, milk, serum, Tissue Culture Media samples.