Mouse Interferon gamma ELISPOT Kit (with un-coated plates)
: Abcam
Mouse Interferon gamma ELISPOT kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Mouse Interferon gamma production in a single cell suspension.
- Species reactivity:Mouse
- Sample type:Suspension cells
- Form:Uncoated
- Antigen name:Interferon gamma
- Storage temperature:+4 ℃
- Processing time:Multiple steps
- Regulatory status:RUO
Frequently Bought Together