Order Entry
Belgium
ContactUsLinkComponent
Anti-MCPyV_gp3 large T antigen Mouse Monoclonal Antibody [clone: Ab3]
Anti-MCPyV_gp3 large T antigen Mouse Monoclonal Antibody [clone: Ab3]
Catalog # ABCAAB269582-100
Supplier:  Abcam
undefined
Anti-MCPyV_gp3 large T antigen Mouse Monoclonal Antibody [clone: Ab3]
Catalog # ABCAAB269582-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

Specifications

About this item

Mouse monoclonal [Ab3] to MCPyV_gp3 large T antigen - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MCPyV_gp3 large T antigen
Clonality: Monoclonal
Clone: Ab3
Conjugation:
Epitope: The epitope for monoclonal Ab3 is between amino acid residues 79 and 260 of MCPyV large T antigen. Ab3 does not bind to MCPyV small T antigen (PubMed ID: 23114601)
Host: Mouse
Isotype: IgG1
Reactivity: