Order Entry
Belgium
ContactUsLinkComponent
Anti-Pan Trk Rabbit Monoclonal Antibody [clone: EPR17341-4]
Anti-Pan Trk Rabbit Monoclonal Antibody [clone: EPR17341-4]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Pan Trk Rabbit Monoclonal Antibody [clone: EPR17341-4]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB246551-100
  • ABCAAB246551-1ML
  • Antigen symbol:
  • Pan Trk
  • Pan Trk
  • Antigen name:
  • TRKB
  • TRKB
  • Antigen synonyms:
  • NTRK2|BDNF/NT-3 growth factors receptor|GP145-Neurotrophic tyrosine kinase receptor type 2|TrkB tyrosine kinase|Tropomyosin-related kinase B|Trk-B
  • NTRK2|BDNF/NT-3 growth factors receptor|GP145-Neurotrophic tyrosine kinase receptor type 2|TrkB tyrosine kinase|Tropomyosin-related kinase B|Trk-B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR17341-4
  • EPR17341-4
  • Form:
  • Liquid
  • Liquid

Specifications

Frequently Bought Together