Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Ornithine Decarboxylase/ODC Mouse Monoclonal Antibody [clone: ODC1/485]
  ABCAAB212690-100
 :  Abcam
undefined
Anti-Ornithine Decarboxylase/ODC Mouse Monoclonal Antibody [clone: ODC1/485]
  ABCAAB212690-100
 :  Abcam

 

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

 

 

Mouse monoclonal [ODC1/485] to Ornithine Decarboxylase/ODC - BSA and Azide free.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Ornithine Decarboxylase/ODC
Clonality: Monoclonal
Clone: ODC1/485
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse