Anti-Ornithine Carbamoyltransferase/OTC Rabbit Monoclonal Antibody [clone: EPR19725]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR19725] to Ornithine Carbamoyltransferase/OTC.
Type: Primary
Antigen: Ornithine Carbamoyltransferase/OTC
Clonality: Monoclonal
Clone: EPR19725
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together