- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Mouse
- Target protein:Cysteinyl Leukotrienes C4
- Description:Mouse Cysteinyl Leukotrienes C4 ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Mouse Cysteinyl Leukotrienes C4 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Cysteinyl Leukotrienes C4. No significant cross-reactivity or interference between Cysteinyl Leukotrienes C4 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:2 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Sample volume:50 μl
- Sensitivity:9,375 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Mouse Cysteinyl Leukotrienes C4 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Cysteinyl Leukotrienes C4 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse Cysteinyl Leukotrienes C4 ELISA Kit employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Cysteinyl Leukotrienes C4 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Cysteinyl Leukotrienes C4 antigen. During the incubation, Cysteinyl Leukotrienes C4 present in the samples or standards competes with the fixed amount of immobilised Cysteinyl Leukotrienes C4 for binding sites on the Biotinylated Anti-Cysteinyl Leukotrienes C4 Antibody. The more Cysteinyl Leukotrienes C4 present in a sample or standard, the less Biotinylated Anti-Cysteinyl Leukotrienes C4 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Cysteinyl Leukotrienes C4 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Cysteinyl Leukotrienes C4 present in each sample or standard. The concentration of Cysteinyl Leukotrienes C4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.