Specifications
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Vitamin B12
- Description:Vitamin B12 ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Vitamin B12 ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Vitamin B12. No significant cross-reactivity or interference between Vitamin B12 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:2 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:50 - 500 pg/ml
- Storage temperature:4 °C
- Sample volume:50 μl
- Sensitivity:25 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Vitamin B12 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Vitamin B12 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Vitamin B12 ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of Vitamin B12 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Vitamin B12 antigen. During the incubation, Vitamin B12 present in the samples or standards competes with the fixed amount of immobilised Vitamin B12 for binding sites on the Biotinylated Anti-Vitamin B12 Antibody. The more Vitamin B12 present in a sample or standard, the less Biotinylated Anti-Vitamin B12 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Vitamin B12 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Vitamin B12 present in each sample or standard. The concentration of Vitamin B12 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.